missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates™ pureGrade™ Strip Plates

Product Code. 11643189 Shop All Brand Products
Change view
Click to view available options
Total Volume (Metric) Well:
350 μL
360 μL
Breakapart:
No
Yes
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Total Volume (Metric) Well Breakapart Unit Size
11643189 350 μL Yes Pack of 100
11633189 360 μL No Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11643189 Supplier BRAND™ Supplier No. 782301
  • ONLINE EXCLUSIVE PRICE
    704.00 €
    676.80 € / Pack of 100
    Save 27.20 €
    3% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Non-treated surface strip plates designed to maximize sample recovery and minimize cross-contamination.

  • Medium binding and non-sterile
  • Transparent with flat bottoms
  • Available with or without grid
  • Free from endotoxins, DNase, DNA, RNase, and non-cytotoxic

With grid: Individual strips can be removed from the plate and reinserted. Strips can also be divided into 8 individual vessels and inserted separately into the frame plate.

Without grid: Individual strips can be removed from the frame and reinserted. Strips are not divisible.

Applications: Ideal for homogeneous assays, screening and storage.

TRUSTED_SUSTAINABILITY

Specifications

Barcode Alphanumeric
Breakapart Yes
Binding Property Untreated
Certified Life Science Quality
Format Strip Plate
Array 12 x 8
Autoclavable Not Autoclavable
Binding Type Medium Binding
Color Clear
Disposable Single-use
Product Type Strip plates
For Use With (Equipment) Microplate readers
Material Polystyrene
Volume (Metric) Well 350 μL
No. of Wells 96
Volume (Metric) Working 350 μL
Sterility Non-sterile
Surface Treatment Untreated
Total Volume (Metric) Well 350 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) Homogeneous assays, screenings, and storage
Includes 96-well strip plate
Quantity 100/Pk.
RNase Free Yes
Target Molecule Immunoglobulin G (IgG)
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.