missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates™ immunoGrade™ Strip Plates

Product Code. 11653189 Shop All Brand Products
Change view
Click to view available options
Total Volume (Metric) Well:
350 μL
360 μL
Breakapart:
No
Yes
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Total Volume (Metric) Well Breakapart Unit Size
11653189 360 μL No Pack of 100
11663189 350 μL Yes Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11653189 Supplier BRAND™ Supplier No. 782305
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-binding plates with optimized surfaces for the immobilization of IgG High-binding plates, optimized for immobilizing IgG and molecules with both hydrophilic and hydrophobic regions, deliver reliable performance for a range of assays.

  • High-binding, non-sterile plates
  • Constructed from high-quality, virgin polystyrene
  • Transparent with a flat bottom
  • Free from endotoxins, DNase, DNA, RNase; non-cytotoxic
  • Available with or without a grid

With grid (Part No. 782306):

  • 12 strips of 8 wells each
  • Individual strips can be removed from the plate and reinserted
  • Strips can also be divided into 8 individual vessels and inserted into the frame plate

Without grid (Part No. 782305):

  • 12 strips of 8 wells each
  • Individual strips can be removed from the frame and reinserted
  • Strips are not divisible

Applications:

Designed for solid phase immunoassays, ELISA, RIA, FIA and other immune testing applications

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Breakapart No
Binding Property immunoGrade
Certified DNA, DNase, RNase, Pyrogen free
Format Strip Plate
Array 1 x 8
Autoclavable Not Autoclavable
Packaging Bagged
Binding Type High Binding
Color Clear
Disposable Single-use
Product Type 96-well strip plate
For Use With (Equipment) Microplate readers, gamma counters, fluorescence spectrophotometers
Material Polystyrene
Volume (Metric) Well 360 μL
No. of Wells 8
Volume (Metric) Working 50 to 320 μL
Sterility Non-sterile
Surface Treatment Immunological
Total Volume (Metric) Well 360 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) Solid-phase immunoassay, immune testing applications
Includes Strip plates
Quantity 100/Pk.
RNase Free Yes
Target Molecule Proteins or antibodies
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.