missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4EBP1 Antibody (9E12D9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60514-0.025ml
This item is not returnable.
View return policy
Description
4EBP1 Monoclonal antibody specifically detects 4EBP1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| 4EBP1 | |
| Monoclonal | |
| Unconjugated | |
| Ascites | |
| 4E-BP1, eIF4E-binding protein 1, eukaryotic translation initiation factor 4E binding protein 1,4EBP1,4E-BP1BP-1, eukaryotic translation initiation factor 4E-binding protein 1, PHAS-IMGC4316, Phosphorylated heat- and acid-stable protein regulated by insulin 1 | |
| Purified recombinant fragment of 4EBP1 expressed in E. Coli. | |
| 0.025 mL | |
| Cancer, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, mTOR Pathway, Stem Cell Markers | |
| 1978 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 9E12D9 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Q13541 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction