missing translation for 'onlineSavingsMsg'
Learn More

5-(1-(Hydroxymethyl)cyclopropyl)-2,2-dimethyl-1,3-dioxolan-4-one, TRC

Product Code. 30393974
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393974 5 mg 5mg
30389857 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30393974 Supplier Toronto Research Chemicals Supplier No. TRCH9465405MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(1-(Hydroxymethyl)cyclopropyl)-2,2-dimethyl-1,3-dioxolan-4-one
Analyte or Component Names 5-(1-(Hydroxymethyl)cyclopropyl)-2,2-dimethyl-1,3-dioxolan-4-one
Shipping Condition Room Temperature
Molecular Formula C9 H14 O4
InChI Formula InChI=1S/C9H14O4/c1-8(2)12-6(7(11)13-8)9(5-10)3-4-9/h6,10H,3-5H2,1-2H3
Solution Type Neat
SMILES CC1(C)OC(C(=O)O1)C2(CO)CC2
IUPAC Name 5-[1-(hydroxymethyl)cyclopropyl]-2,2-dimethyl-1,3-dioxolan-4-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 186.205
Quantity 5 mg
Formula Weight 186.089
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.