missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Bromophenyl)isoxazole, ≥95%, Thermo Scientific™

Product Code. 10526133
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10526133 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10526133 Supplier Thermo Scientific Maybridge Supplier No. CC20510DA
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 7064-33-7
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C31H38N4O4S2
MDL Number MFCD02183531
Quantity 1 g
Synonym 5-3-bromophenyl isoxazole, 5-3-bromophenyl-1,2-oxazole, isoxazole,5-3-bromophenyl, zlchem 1341, acmc-209ofi
InChI Key ALUPLTWVMIXEQH-DICXZTSXNA-N
SMILES CCCN1C(=O)C(C#N)=C(C)C(\C=C2/SC(=S)N(CCC3=CC=C(OC)C(OC)=C3)C2=O)=C1N1CC(C)CC(C)C1
IUPAC Name 5-(3-bromophenyl)-1,2-oxazole
Molecular Weight (g/mol) 594.79
PubChem CID 2735607
Formula Weight 224.06
Percent Purity 95%
Chemical Name or Material 5-(3-Bromophenyl)isoxazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.