missing translation for 'onlineSavingsMsg'
Learn More

5-[3-(Dimethylamino)propylidene]dibenzosuberane hydrochloride, 98%

Product Code. 15490457
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490457 100 g 100g
15480457 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490457 Supplier Thermo Scientific Alfa Aesar Supplier No. J65596.22
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A tricyclic serotnonin reuptake inhibitor | CAS: 549-18-8 | C20H24ClN | 313.869 g/mol

A tricyclic serotnonin reuptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 549-18-8
Molecular Formula C20H24ClN
Molecular Weight (g/mol) 313.869
MDL Number MFCD00012537
InChI Key KFYRPLNVJVHZGT-UHFFFAOYSA-N
Synonym amitriptyline hydrochloride, amitriptyline hcl, annoyltin, tryptizol, domical, elavil hydrochloride, lentizol, saroten, amavil, ami-anelun
PubChem CID 11065
IUPAC Name 3-(5,6-dihydrodibenzo[2,1-b
SMILES CN(C)CCC=C1C2=CC=CC=C2CCC3=CC=CC=C31.Cl

Specifications

Melting Point 196°C to 197°C
Color White
Quantity 100 g
UN Number UN2811
Merck Index 14,487
Formula Weight 313.86
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-[3-(Dimethylamino)propylidene]dibenzosuberane hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.