missing translation for 'onlineSavingsMsg'
Learn More

5-[4’-Bromomethyl-(1,1’-biphenyl)-2-yl]-2-triphenylmethyltetrazole, TRC

Product Code. 30134016
Change view
Click to view available options
Quantity:
1 g
2.5 g
10 g
Unit Size:
10g
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134016 10 g 10g
30132685 2.5 g 2.5g
30005878 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134016 Supplier Toronto Research Chemicals Supplier No. TRCB68531010G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133051-88-4
Molecular Formula C33 H25 Br N4
Molecular Weight (g/mol) 557.48
Synonym 5-[4'-(Bromomethyl)[1,1'-biphenyl]-2-yl]-2-(triphenylmethyl)-2H-tetrazole, 5-[4'-(Bromomethyl)biphenyl-2-yl]-2-trityl-2H-tetrazole, [2'-(2-Triphenylmethyl-2H-tetrazol-5-yl)biphenyl-4-yl]methyl bromide
IUPAC Name 5-[2-[4-(bromomethyl)phenyl]phenyl]-2-trityltetrazole
SMILES BrCc1ccc(cc1)c2ccccc2c3nnn(n3)C(c4ccccc4)(c5ccccc5)c6ccccc6

Specifications

Quantity 10 g
InChI Formula InChI=1S/C33H25BrN4/c34-24-25-20-22-26(23-21-25)30-18-10-11-19-31(30)32-35-37-38(36-32)33(27-12-4-1-5-13-27,28-14-6-2-7-15-28)29-16-8-3-9-17-29/h1-23H,24H2
Formula Weight 556.1263
Chemical Name or Material 5-[4'-Bromomethyl-(1,1'-biphenyl)-2-yl]-2-triphenylmethyltetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.