missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Iodophenyl)isoxazole, 97%, Thermo Scientific™

Product Code. 10141062
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10141062 250 mg 250mg
10782463 1 g 1g
10381945 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10141062 Supplier Thermo Scientific Maybridge Supplier No. CC12911CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 160377-48-0
Molecular Formula C9H6INO
Molecular Weight (g/mol) 271.057
MDL Number MFCD02681898
InChI Key TYNWYXFACGRXMG-UHFFFAOYSA-N
Synonym 5-4-iodophenyl isoxazole, 5-4-iodophenyl-1,2-oxazole, isoxazole,5-4-iodophenyl
PubChem CID 2776370
IUPAC Name 5-(4-iodophenyl)-1,2-oxazole
SMILES C1=CC(=CC=C1C2=CC=NO2)I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 271.05
Percent Purity 97%
Chemical Name or Material 5-(4-Iodophenyl)isoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.