missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Methoxyphenyl)-1,3-oxazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10782017
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10782017 5 g 5g
10181262 250 mg 250mg
10427242 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10782017 Supplier Thermo Scientific Maybridge Supplier No. MO00452DE
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 89205-07-2
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.196
MDL Number MFCD03644147
InChI Key XVXCIDWYZMBWJU-UHFFFAOYSA-N
Synonym 5-4-methoxyphenyl-1,3-oxazole-4-carboxylic acid, 5-4-methoxyphenyl oxazole-4-carboxylic acid, 5-4-methoxyphenyl-1,3-oxazole-4-carboxylicacid, 4-oxazolecarboxylicacid, 5-4-methoxyphenyl, 5-4-methoxy-phenyl-oxazole-4-carboxylic acid, maybridge3_004367, acmc-20c54g, 5-4-methoxyphenyl-4-oxazolecarboxylic acid, 5-4-methoxyphenyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 5-4-methoxyphenyl
PubChem CID 2779782
IUPAC Name 5-(4-methoxyphenyl)-1,3-oxazole-4-carboxylic acid
SMILES COC1=CC=C(C=C1)C2=C(N=CO2)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 219.2
Percent Purity 97%
Chemical Name or Material 5-(4-Methoxyphenyl)-1, 3-oxazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.