missing translation for 'onlineSavingsMsg'
Learn More

5,5,7,7-Tetramethyl-1,5,6,7-tetrahydrocyclopenta[f]indazol-4-amine, Thermo Scientific™

Product Code. 10087929
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10087929 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10087929 Supplier Thermo Scientific Maybridge Supplier No. MAY00301DA
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1174064-61-9
Molecular Formula C14H19N3
MDL Number MFCD11109513
Quantity 1 g
Synonym 5,5,7,7-tetramethyl-1,5,6,7-tetrahydrocyclopenta f indazol-4-amine, 5,5,7,7-tetramethyl-1h,6h-cyclopenta f indazol-4-amine, 1,5,6,7-tetrahydro-5,5,7,7-tetramethylcyclopenta f indazol-4-amine, 5,5,7,7-tetramethyl-1,5,6,7-tetrahydrocyclopenta-f indazol-4-amine, 5,5,7,7-tetramethyl-1h,5h,6h,7h-cyclopenta f indazol-4-amine, 5,5,7,7-tetramethyl-5,6,7-trihydro-1h-cyclopenta 2,1-f indazole-4-ylamine
InChI Key IOJZKUQKQPKIQK-UHFFFAOYSA-N
SMILES CC1(C)CC(C)(C)C2=C1C=C1NN=CC1=C2N
IUPAC Name 5,5,7,7-tetramethyl-1H,5H,6H,7H-cyclopenta[f]indazol-4-amine
Molecular Weight (g/mol) 229.33
PubChem CID 45594317
Formula Weight 229.33
Chemical Name or Material 5,5,7,7-Tetramethyl-1,5,6,7-tetrahydrocyclopenta[f]indazol-4-amine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.