missing translation for 'onlineSavingsMsg'
Learn More

5,5,8,8-Tetramethyl-5,6,7,8-tetrahydro-2-naphthalenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10484183
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10484183 250 mg Amber Glass Bottle 250mg
10671865 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10484183 Supplier Thermo Scientific Maybridge Supplier No. CC27103CB
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 132392-26-8
Molecular Formula C14H19ClO2S
Molecular Weight (g/mol) 286.814
MDL Number MFCD04115384
InChI Key BEAYCJSJSMOLFG-UHFFFAOYSA-N
Synonym 5,5,8,8-tetramethyl-5,6,7,8-tetrahydro-2-naphthalenesulfonyl chloride, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene-2-sulfonyl chloride, 5,6,7,8-tetrahydro-5,5,8,8-tetramethylnaphthalene-2-sulfonyl chloride
PubChem CID 2795467
IUPAC Name 5,5,8,8-tetramethyl-6,7-dihydronaphthalene-2-sulfonyl chloride
SMILES CC1(CCC(C2=C1C=CC(=C2)S(=O)(=O)Cl)(C)C)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 286.82
Percent Purity 97%
Chemical Name or Material 5,5,8,8-Tetramethyl-5,6,7,8-tetrahydro-2-naphthalenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.