missing translation for 'onlineSavingsMsg'
Learn More

5,7,12,14-Pentacenetetrone, 95%

Product Code. 11440937
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440937 5 g 5g
11430937 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440937 Supplier Thermo Scientific Alfa Aesar Supplier No. B23321.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23912-79-0 | C22H10O4 | 338.318 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23912-79-0
Molecular Formula C22H10O4
Molecular Weight (g/mol) 338.318
MDL Number MFCD00075209
InChI Key YZOGOBWHTVNKGA-UHFFFAOYSA-N
Synonym 5,7,12,14-pentacenetetrone, 2.3-phthalylanthrachinon, pentacene-5,7,12,14-tetraone, 5,7,12,14-tetraoxo-5,7,12,14-tetrahydropentacene, 5,7,12,14-tetrahydropentacene-5,7,12,14-tetrone
PubChem CID 4733
IUPAC Name pentacene-5,7,12,14-tetrone
SMILES C1=CC=C2C(=C1)C(=O)C3=CC4=C(C=C3C2=O)C(=O)C5=CC=CC=C5C4=O

Specifications

Quantity 5 g
Formula Weight 338.32
Percent Purity ≥95%
Chemical Name or Material 5,7,12,14-Pentacenetetrone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.