missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dihydroxy 4-methylcoumarin, TRC

Product Code. 30008178
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008178 500 mg 500mg
30012034 250 mg 250mg
30010698 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008178 Supplier Toronto Research Chemicals Supplier No. TRCD449171500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2107-76-8
Molecular Formula C10 H8 O4
Molecular Weight (g/mol) 192.17
Synonym 2H-1-Benzopyran-2-one, 5,7-dihydroxy-4-methyl-, Coumarin, 5,7-dihydroxy-4-methyl- (7CI,8CI), 4-Methyl-5,7-dihydroxycoumarin, 5,7-Dihydroxy-4-methyl-chromen-2-one, 5,7-Dihydroxy-4-methylcoumarin, NSC 5302
IUPAC Name 5,7-dihydroxy-4-methylchromen-2-one
SMILES CC1=CC(=O)Oc2cc(O)cc(O)c12

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H8O4/c1-5-2-9(13)14-8-4-6(11)3-7(12)10(5)8/h2-4,11-12H,1H3
Formula Weight 192.0423
Chemical Name or Material 5,7-Dihydroxy-4-Methylcoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.