missing translation for 'onlineSavingsMsg'
Learn More

5-Acetamido-2-aminobenzoic acid, 97%

Product Code. 11327656
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327656 5 g 5g
11337656 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327656 Supplier Thermo Scientific Alfa Aesar Supplier No. L11256.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 50670-83-2 | C9H10N2O3 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50670-83-2
Molecular Formula C9H10N2O3
Molecular Weight (g/mol) 194.19
MDL Number MFCD00060120
InChI Key GSOHXJQXAKNJES-UHFFFAOYSA-N
Synonym 5-acetamidoanthranilic acid, 2-amino-5-acetamidobenzoic acid, benzoic acid, 5-acetylamino-2-amino, 5-acetylamino-2-aminobenzoic acid, 5-acetamino-2-aminobenzoic acid, 5-acetamidoanthranilicacid, acmc-209kp2, 5-acetylaminoanthanilic acid, 5-acetamido-2-aminobenzoicacid, 2-amino-5-acetaminobenzoic acid
PubChem CID 170890
IUPAC Name 5-acetamido-2-aminobenzoic acid
SMILES CC(=O)NC1=CC(=C(C=C1)N)C(=O)O

Specifications

Melting Point ∽230°C (decomposition)
Quantity 5 g
Beilstein 2726882
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 5-Acetamido-2-aminobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.