missing translation for 'onlineSavingsMsg'
Learn More

5-Acetamido-2-aminobenzoic acid, 97%

Product Code. p-7045586
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11327656

Brand: Thermo Scientific Alfa Aesar L11256.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50670-83-2 | C9H10N2O3 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50670-83-2
Molecular Formula C9H10N2O3
Molecular Weight (g/mol) 194.19
MDL Number MFCD00060120
InChI Key GSOHXJQXAKNJES-UHFFFAOYSA-N
Synonym 5-acetamidoanthranilic acid, 2-amino-5-acetamidobenzoic acid, benzoic acid, 5-acetylamino-2-amino, 5-acetylamino-2-aminobenzoic acid, 5-acetamino-2-aminobenzoic acid, 5-acetamidoanthranilicacid, acmc-209kp2, 5-acetylaminoanthanilic acid, 5-acetamido-2-aminobenzoicacid, 2-amino-5-acetaminobenzoic acid
PubChem CID 170890
IUPAC Name 5-acetamido-2-aminobenzoic acid
SMILES CC(=O)NC1=CC(=C(C=C1)N)C(=O)O

Specifications

Melting Point ∽230°C (decomposition)
Quantity 5 g
Beilstein 2726882
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 5-Acetamido-2-aminobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.