missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1,2,4-thiadiazole, 97%

Product Code. 15274784 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
15274784 1 g Glass bottle 1g
15284784 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 15274784

Brand: Thermo Scientific Acros 449870010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7552-07-0 | C2H3N3S | 101.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7552-07-0
Molecular Formula C2H3N3S
Molecular Weight (g/mol) 101.13
InChI Key VJHTZTZXOKVQRN-UHFFFAOYSA-N
Synonym 5-amino-1,2,4-thiadiazole, 1,2,4-thiadiazol-5-ylamine, 1,2,4-thiadiazole-5-ylamine, 1,2,4-thiadiazole, 5-amino, pubchem11136, acmc-1blyp, 1,2,4-thiadiazole-5-amine, 1,4-thiadiazole, 5-amino, 2-amino-1,3,5-thiadiazole, 5-imino-1,2,4-thiadiazole
PubChem CID 290274
IUPAC Name 1,2,4-thiadiazol-5-amine
SMILES C1=NSC(=N1)N

Specifications

Boiling Point 112.0°C to 121.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 101.13
Percent Purity 97%
Chemical Name or Material 5-Amino-1, 2, 4-thiadiazole
compliance-icons
Product Identifier
  • 5-Amino-1,2,4-thiadiazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.