missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2,4-difluorophenol, 98%, Thermo Scientific™

Product Code. 15485197
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485197 1 g 1g
15495197 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15485197 Fornitore Thermo Scientific Alfa Aesar N. del fornitore H32479.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 113512-71-3
Molecular Formula C6H5F2NO
Molecular Weight (g/mol) 145.109
MDL Number MFCD04973718
InChI Key CXADAFQCFSVRGN-UHFFFAOYSA-N
Synonym 5-amino-2,4-difluorobenzenol, 2,4-difluoro-5-hydroxyaniline, phenol, 5-amino-2,4-difluoro, aminodifluorobenzenol, acmc-20e9pc, 2,4-difluoro-5-aminophenol, 2,4-difluoro-5-amino-phenol
PubChem CID 2782999
IUPAC Name 5-amino-2,4-difluorophenol
SMILES C1=C(C(=CC(=C1O)F)F)N

Specifica

Melting Point ∽177°C (decomposition)
Quantity 1 g
Formula Weight 145.11
Percent Purity 98%
Chemical Name or Material 5-Amino-2,4-difluorophenol
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.