missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-nitrobenzoic acid, 95%

Product Code. 11392937
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392937 25 g 25g
11372937 1 g 1g
11382937 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11392937 Supplier Thermo Scientific Alfa Aesar Supplier No. L16225.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13280-60-9 | C7H6N2O4 | 182.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13280-60-9
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.135
MDL Number MFCD00035748
InChI Key KZZWQCKYLNIOBT-UHFFFAOYSA-N
Synonym 5-amino-2-nitrobenzoic acid, 2-nitro-5-aminobenzoic acid, benzoic acid, 5-amino-2-nitro, 3-carboxy-4-nitroaniline, 5-amino-2-nitro-benzoic acid, pubchem4983, acmc-1bxcb, intermediates-zcf02020, 5-amino-2-nitrobenzoicacid, 6-nitro-m-anthranilic acid
PubChem CID 83298
SMILES C1=CC(=C(C=C1N)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point ∽240°C (decomposition)
Quantity 25 g
Beilstein 2107512
Formula Weight 182.14
Percent Purity 95%
Chemical Name or Material 5-Amino-2-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.