missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3H-imidazole-4-Carboxamide, MedChemExpress

Product Code. 16431929
Encompass_Preferred
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 16431929

Brand: Medchem Express HY41461

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 5-Amino-3H-imidazole-4-Carboxamide (AICA) is an important precursor for the synthesis of purines in general and of the nucleobases adenine and guanine in particular.

5-Amino-3H-imidazole-4-Carboxamide (AICA) is an important precursor for the synthesis of purines in general and of the nucleobases adenine and guanine in particular.

Specifications

Chemical Name or Material 5-Amino-3H-imidazole-4-Carboxamide
Physical Form Solid
CAS 360-97-4
Color Light Khaki
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C4H6N4O
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym 5-Aminoimidazole-4-carboxamide AICA
Solubility Information DMSO : 100 mg/mL (792.90 mM; Need ultrasonic) ∣H2O : 20 mg/mL (158.58 mM; Need ultrasonic)
SMILES O=C(C1=C(N)NC=N1)N
Molecular Weight (g/mol) 126.12
Formula Weight 126.12
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.