missing translation for 'onlineSavingsMsg'
Learn More

5-[[Amino(imino)methyl]amino]-2-(benzoylamino)pentanoic acid, 97%, Thermo Scientific™

Product Code. 11396925
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11396925 25 g 25g
10591312 1 g 1g
10406672 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396925 Supplier Thermo Scientific Maybridge Supplier No. SB01487EB
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 6453-58-3
Molecular Formula C13H18N4O3
Molecular Weight (g/mol) 278.312
MDL Number MFCD00063011
InChI Key RSYYQCDERUOEFI-UHFFFAOYSA-N
Synonym 5-amino imino methyl amino-2-benzoylamino pentanoic acid, 5-carbamimidamido-2-phenylformamido pentanoic acid, n2-benzoylarginine, l-arginine, n2-benzoyl, benzoyl-dl-arginine, n-?-benzoyl-l-arginine, 2-benzamido-5-guanidinopentanoic acid, 2-benzoylamino-5-guanidinovaleric acid, arginine,n2-benzoyl-,monohydrochloride 9ci, 2-benzamido-5-diaminomethylideneamino pentanoic acid
PubChem CID 273355
IUPAC Name 2-benzamido-5-(diaminomethylideneamino)pentanoic acid
SMILES C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula C6H5C(O)NHCH(CO2H)CH2CH2CH2NHC(NH)NH2
Quantity 25 g
Formula Weight 278.31
Percent Purity 97%
Chemical Name or Material 5-[[Amino(imino)methyl]amino]-2-(benzoylamino)pentanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.