missing translation for 'onlineSavingsMsg'
Learn More

5-Aminonaphthalene-2-sulfonic acid, 97%

Product Code. 11322026
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11322026

Brand: Thermo Scientific Alfa Aesar L03099.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119-79-9 | C10H9NO3S | 223.246 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-79-9
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.246
MDL Number MFCD00004030
InChI Key UWPJYQYRSWYIGZ-UHFFFAOYSA-N
Synonym 5-amino-2-naphthalenesulfonic acid, 1,6-cleve's acid, 1-aminonaphthalene-6-sulfonic acid, 2-naphthalenesulfonic acid, 5-amino, 1-naphthylamine-6-sulfonic acid, kyselina cleve, cleve's acid-1,6, 1-amino-6-sulfonaphthalene, 5-naphthylamine-2-sulfonic acid, cleve's beta-acid
PubChem CID 8408
ChEBI CHEBI:44188
IUPAC Name 5-aminonaphthalene-2-sulfonic acid
SMILES C1=CC2=C(C=CC(=C2)S(=O)(=O)O)C(=C1)N

Specifications

Melting Point >300°C
Odor Odorless
Quantity 25 g
Beilstein 1819887
Merck Index 14,2350
Formula Weight 223.25
Percent Purity 97%
Chemical Name or Material 5-Aminonaphthalene-2-sulfonic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.