missing translation for 'onlineSavingsMsg'
Learn More

5-Azacytidine, 98%, Thermo Scientific™

Causes DNA demethylation or hemi-demethylation; a powerful mutagen

Brand:  Thermo Scientific Alfa Aesar J64159.03


Quantity 1g

 View more versions of this product

Product Code. 15426239

  • ONLINE EXCLUSIVE PRICE
    103.00 €
    101.00 € / 1g
    Save 2.00 €
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Azacytidine, its incorporation into RNA alters RNA synthesis and processing, and results in inhibition of protein synthesis. It has been used as a cancer chemotherapeutic agent. It is a powerful bacteriostatic, antitumor, and mutagenic agent; it also exhibits immunosuppressive, antimitotic, radioprotective, and virostatic effects.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

320-67-2
244.207
NMUSYJAQQFHJEW-KVTDHHQDSA-N
9444
4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,3,5-triazin-2-one
C8H12N4O5
MFCD00006539
5-azacytidine, azacitidine, ladakamycin, azacytidine, vidaza, mylosar, azacitidina, azacitidinum, 5-azacitidine, azacitidinum inn-latin
CHEBI:2038
C1=NC(=NC(=O)N1C2C(C(C(O2)CO)O)O)N

Specifications

320-67-2
MFCD00006539
620461
5-azacytidine, azacitidine, ladakamycin, azacytidine, vidaza, mylosar, azacitidina, azacitidinum, 5-azacitidine, azacitidinum inn-latin
NMUSYJAQQFHJEW-KVTDHHQDSA-N
4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,3,5-triazin-2-one
9444
244.2
5-Azacytidine
C8H12N4O5
1g
14,890
Soluble in water.
C1=NC(=NC(=O)N1C2C(C(C(O2)CO)O)O)N
244.207
CHEBI:2038
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Azacytidine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B

Hazard Statement
  • H302-Harmful if swallowed.
  • H340-May cause genetic defects.
  • H350-May cause cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : 5-Azacytidine

SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.