missing translation for 'onlineSavingsMsg'
Learn More

5-(Benzyloxy)-2-bromo-4-methylaniline, 97%, Thermo Scientific™

Product Code. 10334102
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10334102 1 g Amber Glass Bottle 1g
10314202 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10334102 Supplier Thermo Scientific Maybridge Supplier No. MO00837DA
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 499770-88-6
Molecular Formula C14H14BrNO
Molecular Weight (g/mol) 292.176
MDL Number MFCD04973043
InChI Key FEMSDHJSFUHYSH-UHFFFAOYSA-N
Synonym 5-benzyloxy-2-bromo-4-methylaniline, benzyloxybromomethylaniline, 5-benzyloxy-2-bromo-4-methyl-phenylamine
PubChem CID 7018023
IUPAC Name 2-bromo-4-methyl-5-phenylmethoxyaniline
SMILES CC1=CC(=C(C=C1OCC2=CC=CC=C2)N)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 292.17
Percent Purity 97%
Chemical Name or Material 5-(Benzyloxy)-2-bromo-4-methylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.