missing translation for 'onlineSavingsMsg'
Learn More

5% Biodiesel/95% High Cetane Diesel Fuel, v/v, VHG™

Product Code. 17984296
Change view
Click to view available options
Quantity:
20 mL
50 mL
250 mL
Unit Size:
20mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17984296 250 mL 250mL
17934176 20 mL 20mL
17952506 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17984296 Supplier VHG Labs Supplier No. VHGBDBLEND5P250
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

5% Biodiesel/95% High Cetane Diesel Fuel, v/v
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Biodiesel/Diesel Fuel Blends
Analyte or Component Names Biodiesel/Diesel Fuel Blends
Shipping Condition Room Temperature
Concentration 34820%
IUPAC Name Biodiesel@fs:Diesel Fuel Blends
Solvent or Matrix High Cetane Diesel Fuel/Biodiesel
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.