missing translation for 'onlineSavingsMsg'
Learn More

5-(Boc-amino)-2-chlorobenzoic acid, 98+%

Product Code. 15448947
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448947 5 g 5g
15458947 250 mg 250mg
15438947 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15448947 Supplier Thermo Scientific Alfa Aesar Supplier No. H52308.06
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 503555-96-2 | C12H14ClNO4 | 271.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 503555-96-2
Molecular Formula C12H14ClNO4
Molecular Weight (g/mol) 271.70
MDL Number MFCD02682166
InChI Key QNRXWUBPCIBQMX-UHFFFAOYSA-N
Synonym 5-tert-butoxycarbonyl amino-2-chlorobenzoic acid, n-boc-5-amino-2-chlorobenzoic acid, 5-boc-amino-2-chlorobenzoic acid, 5-tert-butoxy carbonyl amino-2-chlorobenzoic acid, 2-chloro-5-2-methyl-2-propanyl oxy carbonyl amino benzoic acid, benzoic acid, 2-chloro-5-1,1-dimethylethoxy carbonyl amino-9ci
PubChem CID 22291441
IUPAC Name 2-chloro-5-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid
SMILES CC(C)(C)OC(=O)NC1=CC(C(O)=O)=C(Cl)C=C1

Specifications

Quantity 5 g
Formula Weight 271.7
Percent Purity ≥98%
Chemical Name or Material 5-(Boc-amino)-2-chlorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.