missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-[3-(chloromethyl)phenyl]pyrimidine, 97%, Thermo Scientific™

Product Code. 15490508
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490508 5 g 5g
15480806 250 mg 250mg
15480508 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490508 Supplier Thermo Scientific Alfa Aesar Supplier No. H54135.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1100598-50-2
Molecular Formula C11H8BrClN2
Molecular Weight (g/mol) 283.553
MDL Number MFCD20527211
InChI Key KHSZEUJOVUJWKR-UHFFFAOYSA-N
Synonym 5-bromo-2-3-chloromethyl phenyl pyrimidine, 5-bromo-2-3-chloromethylphenyl pyrimidine, 5-bromo-2-3-chloromethyl-phenyl-pyrimidine, pyrimidine, 5-bromo-2-3-chloromethyl phenyl
PubChem CID 59438047
IUPAC Name 5-bromo-2-[3-(chloromethyl)phenyl]pyrimidine
SMILES C1=CC(=CC(=C1)CCl)C2=NC=C(C=N2)Br

Specifications

Quantity 5 g
Formula Weight 283.55
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-[3-(chloromethyl)phenyl]pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.