missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloroaniline, 98%

Product Code. 15491149
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15491149 25 g 25g
15481149 5 g 5g
15401159 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491149 Supplier Thermo Scientific Alfa Aesar Supplier No. H64104.14
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 60811-17-8 | C6H5BrClN | 206.47 g/mol

5-Bromo-2-chloroaniline is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-chloroaniline is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60811-17-8
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.47
MDL Number MFCD06796283
InChI Key UGOLEPGQWYPIBR-UHFFFAOYSA-N
Synonym 2-chloro-5-bromoaniline, 3-bromo-6-chloroaniline, 5-bromo-2-chlorophenylamine, benzenamine, 5-bromo-2-chloro, 5-bromo-2-chloro-aniline, 5-bromo-2-chloro-phenylamine, 5-bromo-2-chloro aniline, pubchem3132, 5-bromo-2-chlorobenzenamine, ksc493o4j
PubChem CID 3830731
IUPAC Name 5-bromo-2-chloroaniline
SMILES NC1=CC(Br)=CC=C1Cl

Specifications

Melting Point 145°C to 150°C
Quantity 25 g
UN Number UN2811
Beilstein 2716274
Solubility Information Slightly soluble in water.
Formula Weight 206.47
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-chloroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.