missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxy-3-methoxybenzaldehyde, 97%, Thermo Scientific™

Product Code. 10694565
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10694565 100 g 100g
10386884 10 g 10g
10684565 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10694565 Supplier Thermo Scientific Maybridge Supplier No. BTB03758FA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5034-74-2
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
MDL Number MFCD00016593
InChI Key MMFKBTPDEVLIOR-UHFFFAOYSA-N
PubChem CID 262238
IUPAC Name 5-bromo-2-hydroxy-3-methoxybenzaldehyde
SMILES COC1=C(C(=CC(=C1)Br)C=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3OC6H2(OH)(Br)CH(O)
Quantity 100 g
Synonym 5-bromo-3-methoxysalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy-3-methoxy, 5-bromo-2-vanillin, acmc-20am5q, 3-methoxy-5-bromosalicylaldehyde, 5bromo-2-hydroxy-3-methoxy-benzaldehyde, 5-bromo-2-hydroxy-3-methoxy-benzaldehyde, benzaldehyde,5-bromo-2-hydroxy-3-methoxy, 5-bromo-2-hydroxy-3-methoxybenzaldehyde
Formula Weight 231.04
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-hydroxy-3-methoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.