missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-iodobenzonitrile, 98+%

Product Code. 11471337
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471337 25 g 25g
11451337 1 g 1g
11461337 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471337 Supplier Thermo Scientific Alfa Aesar Supplier No. B24880.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121554-10-7 | C7H3BrIN | 307.916 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121554-10-7
Molecular Formula C7H3BrIN
Molecular Weight (g/mol) 307.916
MDL Number MFCD00672950
InChI Key JIGKPMMZNMQXDL-UHFFFAOYSA-N
PubChem CID 7020874
IUPAC Name 5-bromo-2-iodobenzonitrile
SMILES C1=CC(=C(C=C1Br)C#N)I

Specifications

Melting Point 120°C to 122°C
Quantity 25 g
UN Number UN3276
Beilstein 4860282
Sensitivity Light sensitive
Synonym benzonitrile, 5-bromo-2-iodo, 5-bromo-2-iodobenzenecarbonitrile, 2-iodo-5-bromobenzonitrile, 2-cyano-5-bromo-1-iodobenzene, 5-bromo-2-iodobenzonitrile, 98+%, pubchem3790, acmc-1cir6, 5-bromo-2-iodo-benzonitrile, ksc493s3b, benzonitrile,5-bromo-2-iodo
Formula Weight 307.92
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-iodobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.