missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-iodotoluene, 98+%

Product Code. 11369739
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369739 25 g 25g
11359739 5 g 5g
11379739 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11369739 Supplier Thermo Scientific Alfa Aesar Supplier No. B21878.14
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 116632-39-4 | C7H6BrI | 296.93 g/mol

5-Bromo-2-iodotoluene is used as a medical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-iodotoluene is used as a medical intermediate and in organic synthesis.

Notes
Light sensitive. Incompatible with Strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116632-39-4
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.93
MDL Number MFCD00060664
InChI Key GHTUADBHTFHMNI-UHFFFAOYSA-N
Synonym 5-bromo-2-iodotoluene, 1-bromo-4-iodo-5-methylbenzene, benzene, 4-bromo-1-iodo-2-methyl, bromoiodotoluene, pubchem3793, 2-iodo-5-bromo-toluene, 5-bromo-2-iodo toluene, 5-bromo-2-iodotoluene,, 4-bromo-2-methyliodobenzene, 4-bromo-2-methyl-iodobenzene
PubChem CID 2724601
IUPAC Name 4-bromo-1-iodo-2-methylbenzene
SMILES CC1=CC(Br)=CC=C1I

Specifications

Density 2.139
Boiling Point 262°C to 265°C
Flash Point >110°C (230°F)
Refractive Index 1.6515
Quantity 25 g
Beilstein 2325157
Sensitivity Light sensitive
Formula Weight 296.93
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-iodotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.