missing translation for 'onlineSavingsMsg'
Learn More

5-Carbamoyl-2-fluorobenzeneboronic acid, 95%

Product Code. 15419787
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419787 1 g 1g
15429787 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419787 Supplier Thermo Scientific Alfa Aesar Supplier No. H53010.03
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 874289-39-1 | C7H7BFNO3 | 182.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874289-39-1
Molecular Formula C7H7BFNO3
Molecular Weight (g/mol) 182.95
MDL Number MFCD08436055
InChI Key NDVPMVJEURRBQM-UHFFFAOYSA-N
Synonym 3-borono-4-fluorobenzamide, 5-carbamoyl-2-fluorophenyl boronic acid, 5-carbamoyl-2-fluorobenzeneboronic acid, 5-carbamoyl-2-fluorophenyboronic acid, acmc-209qmh, 2-fluoro-5-carbamoylphenylboronic acid, 5-carbamoyl-2-fluorophenyl boronicacid, 5-carbamoyl-2-fluoro-phenyl boronic acid, 5-carbamoyl-2-fluorobenzeneboronicacid
PubChem CID 44717478
IUPAC Name (5-carbamoyl-2-fluorophenyl)boronic acid
SMILES NC(=O)C1=CC(B(O)O)=C(F)C=C1

Specifications

Melting Point 192°C to 194°C
Quantity 1 g
Formula Weight 182.95
Percent Purity 95%
Chemical Name or Material 5-Carbamoyl-2-fluorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.