missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2,4-dinitrotoluene, 97%

Product Code. 11382627
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382627 5 g 5g
11392627 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382627 Supplier Thermo Scientific Alfa Aesar Supplier No. L11808.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51676-74-5 | C7H5ClN2O4 | 216.577 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51676-74-5
Molecular Formula C7H5ClN2O4
Molecular Weight (g/mol) 216.577
MDL Number MFCD00041235
InChI Key KPDPGZNHKMJEFZ-UHFFFAOYSA-N
Synonym 5-chloro-2,4-dinitrotoluene, toluene, 5-chloro-2,4-dinitro, 1-chloro-5-methyl-2,4-dinitro-benzene, 2-chloro-4-methyl-1,5-dinitrobenzene, acmc-20am4s, 5-chlor-2.4-dinitrotoluol, benzene,1-chloro-5-methyl-2,4-dinitro, 1-chloranyl-5-methyl-2,4-dinitro-benzene
PubChem CID 253802
IUPAC Name 1-chloro-5-methyl-2,4-dinitrobenzene
SMILES CC1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])Cl

Specifications

Melting Point 89°C to 91°C
Quantity 5 g
UN Number UN2811
Beilstein 2506591
Formula Weight 216.58
Percent Purity 97%
Chemical Name or Material 5-Chloro-2,4-dinitrotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.