missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2-fluorophenyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10782167
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10782167 5 g 5g
10762123 1 g 1g
10497432 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10782167 Supplier Thermo Scientific Maybridge Supplier No. SB01971DE
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 247170-25-8
Molecular Formula C7H3ClFNS
Molecular Weight (g/mol) 187.616
MDL Number MFCD01313224
InChI Key RUASREJPMXNHLF-UHFFFAOYSA-N
Synonym 5-chloro-2-fluorophenyl isothiocyanate, 5-chloro-2-fluorophenylisothiocyanate, 3-chloro-6-fluorophenylisothiocyanate, 5-chloro-2-fluorobenzenisothiocyanate, benzene,4-chloro-1-fluoro-2-isothiocyanato, 4-chloranyl-1-fluoranyl-2-isothiocyanato-benzene, benzene, 4-chloro-1-fluoro-2-isothiocyanato-9ci
PubChem CID 2736579
IUPAC Name 4-chloro-1-fluoro-2-isothiocyanatobenzene
SMILES C1=CC(=C(C=C1Cl)N=C=S)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 187.62
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-fluorophenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.