missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxyaniline, 98%

Product Code. p-7021772
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11472367

Brand: Thermo Scientific Alfa Aesar A14244.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-03-4 | C7H8ClNO | 157.597 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-03-4
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.597
MDL Number MFCD00007777
InChI Key WBSMIPLNPSCJFS-UHFFFAOYSA-N
Synonym 2-amino-4-chloroanisole, 5-chloro-o-anisidine, benzenamine, 5-chloro-2-methoxy, 2-methoxy-5-chloroaniline, tulabase fast red r, tulabase fast red rc, 4-chloro-2-aminoanisole, o-anisidine, 5-chloro, 5-chloro-2-methoxyphenylamine, p-chloro-o-aminoanisole
PubChem CID 66763
IUPAC Name 5-chloro-2-methoxyaniline
SMILES COC1=C(C=C(C=C1)Cl)N

Specifications

Melting Point 79°C to 84°C
Boiling Point 135°C (8 mmHg)
Odor Characteristic
Quantity 100 g
UN Number UN2233
Beilstein 879161
Formula Weight 157.6
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-methoxyaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.