missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxyphenyl isocyanate, 97%

Product Code. 11400517
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400517 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11400517 Supplier Thermo Scientific Alfa Aesar Supplier No. B22395.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 55440-54-5 | C8H6ClNO2 | 183.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 55440-54-5
Melting Point 58°C to 62°C
Molecular Formula C8H6ClNO2
MDL Number MFCD00013860
Quantity 10 g
UN Number UN2811
Beilstein 2719682
Sensitivity Moisture sensitive
Synonym 5-chloro-2-methoxyphenyl isocyanate, 5-chloro-2-methoxyphenylisocyanate, benzene, 4-chloro-2-isocyanato-1-methoxy, 5-chloro-2-methoxybenzenisocyanate, acmc-20am4l, 2-methoxy-5-chlorophenyl isocyanate, 3-chloro-6-methoxyphenyl isocyanate, 2-methoxy-5-chloro phenyl isocyanate, 5-chloro-2-methoxy phenyl isocyanate, 4-chloro-2-isocyanato-1-methoxy-benzene
InChI Key CDRZZFCLBUGMMB-UHFFFAOYSA-N
SMILES COC1=CC=C(Cl)C=C1N=C=O
IUPAC Name 4-chloro-2-isocyanato-1-methoxybenzene
Molecular Weight (g/mol) 183.59
PubChem CID 144009
Formula Weight 183.6
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-methoxyphenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.