missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylaminobenzophenone, 99%

Product Code. 15488849
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488849 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15488849 Supplier Thermo Scientific Alfa Aesar Supplier No. L06338.09
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1022-13-5 | C14H12ClNO | 245.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1022-13-5
Melting Point 93°C to 98°C
Molecular Formula C14H12ClNO
MDL Number MFCD00008284
Quantity 10 g
Beilstein 882187
Synonym 5-chloro-2-methylamino benzophenone, 5-chloro-2-methylaminobenzophenone, 2-methylamino-5-chlorobenzophenone, 5-chloro-2-methylamino phenyl phenyl methanone, methanone, 5-chloro-2-methylamino phenyl phenyl, unii-d4gd5770pi, benzophenone, 5-chloro-2-methylamino, 2-benzoyl-4-chloro-n-methylaniline, 5-chloro-2-methylamino phenyl phenylmethanone, 5-chloro-2-methylamino phenyl-phenylmethanone
InChI Key WPNMLCMTDCANOZ-UHFFFAOYSA-N
SMILES CNC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1
IUPAC Name [5-chloro-2-(methylamino)phenyl]-phenylmethanone
Molecular Weight (g/mol) 245.71
PubChem CID 13925
Formula Weight 245.71
Percent Purity 99%
Chemical Name or Material 5-Chloro-2-methylaminobenzophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.