missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2-(methylthio)aniline, 95%, Thermo Scientific™

Product Code. 10722493
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10722493 10 g Amber Glass Bottle 10g
10456852 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10722493 Supplier Thermo Scientific Maybridge Supplier No. CD05201EA
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 16423-54-4
Molecular Formula C7H8ClNS
Molecular Weight (g/mol) 173.658
MDL Number MFCD00052816
InChI Key RWUQNIPYCAHKMO-UHFFFAOYSA-N
PubChem CID 2794577
IUPAC Name 5-chloro-2-methylsulfanylaniline
SMILES CSC1=C(C=C(C=C1)Cl)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula NH2C6H3(Cl)SCH3
Quantity 10 g
Synonym 5-chloro-2-methylthio aniline, 5-chloro-2-methylsulfanyl aniline, 5-chloro-2-methylthiophenylamine, 5-chloro-2-methylthio benzenamine, 5-chloro-2-methylsulfanyl-phenylamine, benzenamine,5-chloro-2-methylthio, benzenamine, 5-chloro-2-methylthio
Formula Weight 173.67
Percent Purity 95%
Chemical Name or Material 5-Chloro-2-(methylthio)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.