missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2-nitro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Product Code. 10612695
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10612695 5 g Amber Glass Bottle 5g
10009271 250 mg Amber Glass Bottle 250mg
10282542 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10612695 Supplier Thermo Scientific Maybridge Supplier No. CD04371DE
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 35375-74-7
Molecular Formula C7H4ClF3N2O2
Molecular Weight (g/mol) 240.566
MDL Number MFCD01312277
InChI Key AAMCWLIPRMIPBN-UHFFFAOYSA-N
Synonym 5-chloro-2-nitro-4-trifluoromethyl aniline, 4-amino-2-chloro-5-nitrobenzotrifluoride, 5-chloro-2-nitro-4-trifluoromethyl benzenamine, 3-chloro-6-nitro-4-trifluoromethyl aniline, 5-chloro-2-nitro-4-trifluoromethyl-phenylamine, 5-chloro-2-nitro-4-trifluoromethyl phenylamine, 5-chloro-4-trifluoromethyl-2-nitroaniline, benzenamine, 5-chloro-2-nitro-4-trifluoromethyl, 5-chloro-2-nitro-4-trifluoromethylphenyl amine, pubchem2803
PubChem CID 2773813
IUPAC Name 5-chloro-2-nitro-4-(trifluoromethyl)aniline
SMILES C1=C(C(=CC(=C1[N+](=O)[O-])N)Cl)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 240.57
Percent Purity 97%
Chemical Name or Material 5-chloro-2-nitro-4-(trifluoromethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.