missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-6-nitroisoindoline-1,3-dione, ≥95%, Thermo Scientific™

Product Code. 10476672
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10476672 1 g Amber Glass Bottle 1g
10486672 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10476672 Supplier Thermo Scientific Maybridge Supplier No. TL00400DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6015-57-2
Molecular Formula C8H3ClN2O4
Molecular Weight (g/mol) 226.572
MDL Number MFCD00052331
InChI Key ADLVDYMTBOSDFE-UHFFFAOYSA-N
Synonym 4-chloro-5-nitrophthalimide, 5-chloro-6-nitroisoindoline-1,3-dione, 5-chloro-6-nitro-2h-isoindole-1,3-dione, acmc-1b2sd, 5-chloro-6-nitroindoline-1,3-dione, 5-chloro-6-nitro-isoindole-1,3-dione, 6-chloro-5-nitro-2h-benzo c azoline-1,3-dione, 1h-isoindole-1,3 2h-dione,5-chloro-6-nitro, 6-chloro-5-nitro-2h-benzo c azolidine-1,3-dione, 1h-isoindole-1,3 2h-dione, 5-chloro-6-nitro
PubChem CID 2801129
IUPAC Name 5-chloro-6-nitroisoindole-1,3-dione
SMILES C1=C2C(=CC(=C1[N+](=O)[O-])Cl)C(=O)NC2=O

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 226.57
Percent Purity 95%
Chemical Name or Material 5-Chloro-6-nitroisoindoline-1, 3-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.