missing translation for 'onlineSavingsMsg'
Learn More

5-(Chloromethyl)uracil, 97%

Product Code. 11465460
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11465460

Brand: Thermo Scientific Alfa Aesar 044639.01

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3590-48-5 | C5H5ClN2O2 | 160.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3590-48-5
Molecular Formula C5H5ClN2O2
Molecular Weight (g/mol) 160.56
MDL Number MFCD00218445
InChI Key UCDUBKRXOPMNGH-UHFFFAOYSA-N
Synonym 5-chloromethyl pyrimidine-2,4 1h,3h-dione, 5-chloromethyl-1,3-dihydropyrimidine-2,4-dione, 2,4 1h,3h-pyrimidinedione, 5-chloromethyl, 5-chloromethyluracil, 5-chloromethyl uracil, acmc-1ady4, 5-chloromethyl-2,4-pyrimidinediol, 5-chloromethyl pyrimidine-2,4-diol, 5-chloromethyl uracil 250mg
PubChem CID 236125
SMILES ClCC1=CNC(=O)NC1=O

Specifications

Quantity 0.25 g
IUPAC Name 5-(chloromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Formula Weight 160.56
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5- (Chloromethyl)uracil

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.