missing translation for 'onlineSavingsMsg'
Learn More

5-Chloropyridine-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 15424007
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424007 10 g 10g
15414007 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15424007 Fornitore Thermo Scientific Alfa Aesar N. del fornitore H30923.09
 Restricted Item
Estimated Shipment: 02-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 86873-60-1 | Molecular Formula: C6H4ClNO2 | Molecular Weight: 157.553 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 86873-60-1
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD04114192
InChI Key GJLOKYIYZIOIPN-UHFFFAOYSA-N
Synonym 5-chloropicolinic acid, 5-chloro-2-picolinic acid, 5-chloro-2-pyridinecarboxylic acid, 5-chloro-pyridine-2-carboxylic acid, 5-chloro-2-carboxypyridine, 2-carboxy-5-chloropyridine, 5-chloropyridine-2-carboxylicacid, 2-pyridinecarboxylic acid, 5-chloro, pubchem10379, 5-chloro-picolinic acid
PubChem CID 2762872
IUPAC Name 5-chloropyridine-2-carboxylic acid
SMILES C1=CC(=NC=C1Cl)C(=O)O

Specifica

Melting Point 166°C to 171°C
Quantity 10 g
Formula Weight 157.56
Percent Purity 95%
Chemical Name or Material 5-Chloropyridine-2-carboxylic acid

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.