missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-hydroxybenzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15459457
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459457 250 mg 250mg
15449457 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459457 Supplier Thermo Scientific Alfa Aesar Supplier No. H52716.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 259209-20-6
Molecular Formula C6H6BFO3
Molecular Weight (g/mol) 155.92
MDL Number MFCD03095357
InChI Key TWDQSJDFXUMAOI-UHFFFAOYSA-N
Synonym 5-fluoro-2-hydroxyphenyl boronic acid, 5-fluoro-2-hydroxybenzeneboronic acid, 2-borono-4-fluorophenol, boronic acid, 5-fluoro-2-hydroxyphenyl, 5-fluoro-2-hydroxy phenylboronic acid, boronic acid,b-5-fluoro-2-hydroxyphenyl, pubchem19871, 5-fluoro-2-hydroxy-phenyl boronic acid, acmc-209gna, ablock ab-12-3331
PubChem CID 3772476
IUPAC Name (5-fluoro-2-hydroxyphenyl)boronic acid
SMILES OB(O)C1=C(O)C=CC(F)=C1

Specifications

Melting Point 160°C to 166°C
Quantity 250 mg
Formula Weight 155.92
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-hydroxybenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.