missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylaniline, 99%

Product Code. 11915211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11915211 25 g Glass bottle 25g
11905211 100 g Glass bottle 100g
11925211 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11915211 Supplier Thermo Scientific Acros Supplier No. 119460250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 367-29-3 | C7H8FN | 125.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-29-3
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00007764
InChI Key JLCDTNNLXUMYFQ-UHFFFAOYSA-N
Synonym 2-amino-4-fluorotoluene, 5-fluoro-o-toluidine, 3-fluoro-6-methylaniline, 2-methyl-5-fluoroaniline, benzenamine, 5-fluoro-2-methyl, 5-fluoro-2-methyl-phenylamine, 5-fluoro-2-methyl-aniline, 5-fluoro-2-methylphenylamine, 5-fluor-2-methylanilin
PubChem CID 67774
IUPAC Name 5-fluoro-2-methylaniline
SMILES CC1=CC=C(F)C=C1N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 38.0°C to 40.0°C
Color Purple to Brown
Boiling Point 98.0°C to 100.0°C (15.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(CH3)NH2
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether
Formula Weight 125.15
Percent Purity 99%
Physical Form Crystalline Solid or Liquid
Chemical Name or Material 5-Fluoro-2-methylaniline
Show More Show Less
compliance-icons
Product Identifier
  • 5-Fluoro-2-methylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.