missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylbenzamide, 97%, Thermo Scientific™

Product Code. 10672843
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10672843 1 g 1g
10467632 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10672843 Supplier Thermo Scientific Maybridge Supplier No. TL00714DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-28-1
Molecular Formula C8H8FNO
Molecular Weight (g/mol) 153.156
MDL Number MFCD00205105
InChI Key VPAFHFHYJHZKSR-UHFFFAOYSA-N
Synonym benzamide,5-fluoro-2-methyl, benzamide, 5-fluoro-2-methyl-9ci, 5-fluoro-2-methyl-benzamide, 5-fluoranyl-2-methyl-benzamide, 5-fluoro-2-methylbenzamide, benzamide, 5-fluoro-2-methyl, 2-carbamoyl-4-fluorotoluene, 5-fluoro-o-toluamide
PubChem CID 2737374
IUPAC Name 5-fluoro-2-methylbenzamide
SMILES CC1=C(C=C(C=C1)F)C(=O)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 153.16
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-methylbenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.