missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific™

Product Code. 10027759 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10027759 50 mL AcroSeal™ Glass Bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10027759 Supplier Thermo Scientific Acros Supplier No. 431830500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 186496-59-3, 109-99-9
CAS Min % 8.89, 86.67
CAS Max % 13.33, 91.11
Color Yellow
Density 0.9600g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C7H6BrFMg
Linear Formula F(CH3)C6H3MgBr
MDL Number MFCD01311468
Quantity 50 mL
Specific Gravity 0.96
Synonym bromo 5-fluoro-2-methylphenyl magnesium, magnesium,bromo 5-fluoro-2-methylphenyl, xkmsjpvajjfyfu-uhfffaoysa-m, 5-fluoro-2-methylphenylmagnesium bromide, 2-methyl-5-fluorophenyl magnesium bromide, 5-fluoro-2-methylphenylmagnesium bromide solution, 5-fluoro-2-methylphenylmagnesium bromide, 0.50 m in 2-methf, 5-fluoro-2-methylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 5-fluoro-2-methylphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
Solubility Information Solubility in water: reacts
InChI Key XKMSJPVAJJFYFU-UHFFFAOYSA-M
SMILES CC1=CC=C(F)C=C1[Mg]Br
IUPAC Name magnesium;1-fluoro-4-methylbenzene-5-ide;bromide
Molecular Weight (g/mol) 213.33
PubChem CID 2778349
Formula Weight 213.33
Physical Form Solution
Chemical Name or Material 5-Fluoro-2-methylphenylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 5-Fluoro-2-methylphenylmagnesium bromide, 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.