missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroindole, 98%

Product Code. 13458508 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13458508 25 g Glass bottle 25g
10397521 250 mg Glass bottle 250mg
10183961 1 g Glass bottle 1g
13448508 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13458508 Supplier Thermo Scientific Acros Supplier No. 311920250
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 399-52-0 | C8H6FN | 135.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 399-52-0
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD00005671
InChI Key ODFFPRGJZRXNHZ-UHFFFAOYSA-N
Synonym 5-fluoroindole, 1h-indole, 5-fluoro, 5-fluoro indole, indole, 5-fluoro, 5-fluoro-indole, 5-fluor-1h-indol, 5-fluoroindol, 5-fluoro-indol, 3fuf, pubchem1681
PubChem CID 67861
ChEBI CHEBI:72818
IUPAC Name 5-fluoro-1H-indole
SMILES FC1=CC=C2NC=CC2=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 44.0°C to 48.0°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 135.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoroindole, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.