missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorosalicylic acid, 97%

Product Code. 11480367
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11480367

Brand: Thermo Scientific Alfa Aesar B21751.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 345-16-4 | C7H5FO3 | 156.112 g/mol

5-Fluorosalicylic acid was used as internal standard in the quantification of salicylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluorosalicylic acid was used as internal standard in the quantification of salicylic acid.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 345-16-4
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.112
MDL Number MFCD00002456
InChI Key JWPRICQKUNODPZ-UHFFFAOYSA-N
Synonym 5-fluorosalicylic acid, 2-hydroxy-5-fluorobenzoic acid, benzoic acid, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxybenzoicacid, 5-fluoro-2-hydroxy-benzoic acid, 5fluorosalicylic acid, pubchem1382, 5-fluoro-salicylic acid, 2-carboxy-4-fluorophenol
PubChem CID 67658
IUPAC Name 5-fluoro-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1F)C(=O)O)O

Specifications

Melting Point 178°C to 182°C
Quantity 1 g
Beilstein 2209120
Solubility Information Slightly soluble in water.
Formula Weight 156.11
Percent Purity 97%
Chemical Name or Material 5-Fluorosalicylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.