missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorosalicylic acid, 99%, ACROS Organics™

Product Code. 10722125 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
5g; Glass bottle
This item is not returnable. View return policy

Product Code. 10722125

Brand: Thermo Scientific Acros 166680050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 345-16-4
CAS Min % 98.5
CAS Max % 100.0
Melting Point 179°C to 182°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C7H5FO3
MDL Number MFCD00002456
Quantity 5 g
Synonym 5-fluorosalicylic acid, 2-hydroxy-5-fluorobenzoic acid, benzoic acid, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxybenzoicacid, 5-fluoro-2-hydroxy-benzoic acid, 5fluorosalicylic acid, pubchem1382, 5-fluoro-salicylic acid, 2-carboxy-4-fluorophenol
InChI Key JWPRICQKUNODPZ-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1F)C(=O)O)O
IUPAC Name 5-fluoro-2-hydroxybenzoic acid
Molecular Weight (g/mol) 156.112
PubChem CID 67658
Formula Weight 156.11
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material 5-Fluorosalicylic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.