missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-hmC Antibody (C.15200200), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59177
This item is not returnable.
View return policy
Description
5-hmC Monoclonal specifically detects 5-hmC in Human, Mouse samples. It is validated for ELISA, Dot Blot, Methylated DNA Immunoprecipitation.
Specifications
| 5-hmC | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| HMCES | |
| The exact immunogen is propietary information. | |
| 100 μg | |
| DNA Repair, Epigenetics, Stem Cells | |
| 56941 | |
| Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
| ChIP Assay, Dot Blot | |
| C.15200200 | |
| ELISA 1:500, Dot Blot 2 μg/mL, Methylated DNA Immunoprecipitation 2.5 μg/IP | |
| 5'-hmC, 5-hydroxymethylcytosine | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction