missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-hmC Antibody (RM236), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-61469
This item is not returnable.
View return policy
Description
5-hmC Monoclonal antibody specifically detects 5-hmC in Vertebrate samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot, Methylated DNA Immunoprecipitation
Specifications
| 5-hmC | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1 μg/mL-5 μg/mL, ELISA 0.1 μg/mL - 1 μg/mL, Immunohistochemistry 0.1 μg/mL - 1 μg/mL, Immunocytochemistry/ Immunofluorescence 0.5 μg/mL - 2 μg/mL, Immunohistochemistry-Paraffin, Dot Blot 0.2 μg/mL - 1 μg/mL, Methylated DNA Immunoprecipitation 0.2 μg/mL - 2ug/mL | |
| 5'-hmC, 5-hydroxymethylcytosine | |
| BSA-conjugated 5-hmC | |
| 50 μg | |
| DNA Repair, Epigenetics, Stem Cells | |
| Vertebrates | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| RM236 | |
| Unconjugated | |
| 50% Glycerol/PBS, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction